Merck
  • Association study of fibroblast growth factor 10 (FGF10) polymorphisms with susceptibility to extreme myopia in a Japanese population.
Association study of fibroblast growth factor 10 (FGF10) polymorphisms with susceptibility to extreme myopia in a Japanese population.
Molecular vision (2013-11-23)
Masao Yoshida, Akira Meguro, Eiichi Okada, Naoko Nomura, Nobuhisa Mizuki
ABSTRACT
The fibroblast growth factor 10 (FGF10) gene polymorphism rs339501 was previously reported to be associated with high myopia in a Chinese population. In the present study, we investigated whether FGF10 polymorphisms are associated with extreme myopia in a Japanese population as well. A total of 433 Japanese patients with extreme myopia (≤ -10.00 diopters) and 542 Japanese healthy controls (+1.50 to -1.50 diopters) were recruited. We genotyped seven tagging single-nucleotide polymorphisms (SNPs), including rs339501, in FGF10. We also performed an imputation analysis to evaluate the potential association of ungenotyped FGF10 SNPs, and 34 SNPs were imputed. It was found that rs339501 and rs12517396 exhibited the strongest association with extreme myopia (p=3.9 × 10⁻⁴, corrected p [Pc]=0.0030). A significant association was also observed for rs10462070 (p=6.5 × 10⁻⁴, Pc=0.0059). These three SNPs were in strong linkage disequilibrium (D' ≥0.99, r² ≥0.96). However, the frequency of the A allele of rs339501 was increased in cases compared to controls, which differs from the increased frequency of the G allele in cases in the previous Chinese population. Three FGF10 SNPs in complete linkage disequilibrium--rs339501, rs12517396, and rs10462070--were associated with extreme myopia in the Japanese population, and the risk allele of rs339501 differed from the previous Chinese population. Therefore, these three SNPs may not be an important risk factor for susceptibility to extreme myopia. Further studies are needed to elucidate the possible contribution of the FGF10 region in the development of extreme myopia.
Sigma-Aldrich
Fibroblast Growth Factor-10 human, >97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture