Merck
  • Localization of cytochrome P450 and related enzymes in adult rat testis and downregulation by estradiol and bisphenol A.
Localization of cytochrome P450 and related enzymes in adult rat testis and downregulation by estradiol and bisphenol A.
Toxicological sciences : an official journal of the Society of Toxicology (2014-04-23)
Ravindranath Reddy Gilibili, A Wayne Vogl, Thomas K H Chang, Stelvio M Bandiera
ABSTRACT
There is a growing body of evidence that exposure to endocrine disrupting chemicals and to estrogenic compounds in particular can affect the testis and male fertility. In the present study, the constitutive expression of steroidogenic and non-steroidogenic cytochrome P450 (CYP) and related enzymes in adult rat testis, and their regulation by estradiol and bisphenol A, were investigated. CYP1B1, CYP2A1, NADPH-cytochrome P450 oxidoreductase (POR) and microsomal epoxide hydrolase (mEH) proteins, together with CYP17A1 and 3β-hydroxysteroid dehydrogenase (HSD3B), were detected by immunoblot analysis in testicular microsomes prepared from untreated adult Sprague Dawley rats. In contrast, CYP1A, CYP2B, CYP2E, CYP2D, CYP2C, CYP3A, and CYP4A enzymes were not detected. Immunofluorescence staining of cryosections of perfusion-fixed testes showed that CYP1B1, CYP2A1, CYP17A1, and HSD3B were expressed exclusively or mainly in interstitial cells, whereas mEH and POR protein staining was detected both in interstitial cells and in seminiferous tubules. Testicular CYP1B1 and CYP2A1 protein levels were decreased following treatment of adult rats with estradiol benzoate at 0.004, 0.04, 0.4, or 4 μmol/kg/day or bisphenol A at 400 or 800 μmol/kg/day, for 14 days, whereas expression of HSD3B was unaffected. Testicular CYP17A1, POR, and mEH protein expression was also downregulated at the three highest dosages of estradiol benzoate and at both dosages of bisphenol A. The present study is the first to establish the cellular localization of CYP1B1, mEH, and POR in rat testis and to demonstrate the suppressive effect of bisphenol A on testicular CYP1B1, CYP2A1, mEH, and POR protein levels.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%