Merck
  • Effect of prasugrel pre-treatment strategy in patients undergoing percutaneous coronary intervention for NSTEMI: the ACCOAST-PCI study.
Effect of prasugrel pre-treatment strategy in patients undergoing percutaneous coronary intervention for NSTEMI: the ACCOAST-PCI study.
Journal of the American College of Cardiology (2014-12-20)
Gilles Montalescot, Jean-Philippe Collet, Patrick Ecollan, Leonardo Bolognese, Jurrien Ten Berg, Dariusz Dudek, Christian Hamm, Petr Widimsky, Jean-François Tanguay, Patrick Goldstein, Eileen Brown, Debra L Miller, LeRoy LeNarz, Eric Vicaut
ABSTRACT
After percutaneous coronary intervention (PCI) for non-ST-segment elevation myocardial infarction (NSTEMI), treatment with a P2Y12 antagonist with aspirin is recommended for 1 year. The oral P2Y12 antagonists ticagrelor and prasugrel have higher recommendations than clopidogrel, but it is unknown if administration before the start of PCI is beneficial. In the randomized, double-blind ACCOAST (A Comparison of prasugrel at the time of percutaneous Coronary intervention Or as pre-treatment At the time of diagnosis in patients with non-ST-segment elevation myocardial infarction) trial, 4,033 patients were diagnosed with NSTEMI and 68.7% underwent PCI; 1,394 received pre-treatment with prasugrel (30-mg loading dose), and 1,376 received placebo. At the time of PCI, patients who received pre-treatment with prasugrel received an additional 30-mg dose of prasugrel, and those who received placebo received a 60-mg loading dose of prasugrel. Primary efficacy was a composite of cardiovascular death, myocardial infarction, stroke, urgent revascularization, or glycoprotein IIb/IIIa bailout through 7 days from randomization. Investigators captured the presence of thrombus on initial angiography and during PCI. The incidence of the primary endpoint through 7 days from randomization in the pre-treatment group versus the no pre-treatment group was 13.1% versus 13.1% (p = 0.93). Pre-treatment with prasugrel was not associated with decreases in any ischemic event, including total mortality. Patients with thrombus on angiography had a 3-fold higher incidence of the primary endpoint than patients without thrombus. There was no impact of pre-treatment with prasugrel on the presence of thrombus before PCI or on occurrence of stent thrombosis after PCI. There was a 3-fold increase in all non-coronary artery bypass graft Thrombolysis In Myocardial Infarction (TIMI) major bleeding and a 6-fold increase in non-coronary artery bypass graft life-threatening bleeding with pre-treatment with prasugrel; the same trends persisted in patients who had radial or femoral access even with use of a closure device. These findings support deferring treatment with prasugrel until a decision is made about revascularization in patients with NSTEMI undergoing angiography within 48 h of admission. (A Comparison of prasugrel at the time of percutaneous Coronary intervention Or as pre-treatment At the time of diagnosis in patients with non-ST-segment elevation myocardial infarction [ACCOAST]; NCT01015287).
Sigma-Aldrich
Prasugrel, ≥98% (HPLC)
Sigma-Aldrich
Creatine, anhydrous