Merck
  • The polyserine domain of the lysyl-5 hydroxylase Jmjd6 mediates subnuclear localization.
The polyserine domain of the lysyl-5 hydroxylase Jmjd6 mediates subnuclear localization.
The Biochemical journal (2013-05-22)
Alexander Wolf, Monica Mantri, Astrid Heim, Udo Müller, Erika Fichter, Mukram M Mackeen, Lothar Schermelleh, Gregory Dadie, Heinrich Leonhardt, Catherine Vénien-Bryan, Benedikt M Kessler, Christopher J Schofield, Angelika Böttger
ABSTRACT
Jmjd6 (jumonji-domain-containing protein 6) is an Fe(II)- and 2OG (2-oxoglutarate)-dependent oxygenase that catalyses hydroxylation of lysine residues in proteins involved in pre-mRNA splicing. Jmjd6 plays an essential role in vertebrate embryonic development and has been shown to modulate alternative splicing in response to hypoxic stress. In the present study we show that an alternatively spliced version of Jmjd6 lacking the polyS (polyserine) domain localizes to the nucleolus, predominantly in the fibrillar centre. Jmjd6 with the polyS domain deleted also interacts with nucleolar proteins. Furthermore, co-immunoprecipitation experiments and F2H (fluorescent 2-hybrid) assays demonstrate that Jmjd6 homo-oligomerization occurs in cells. In correlation with the observed variations in the subnuclear distribution of Jmjd6, the structure of Jmjd6 oligomers in vitro changes in the absence of the polyS domain, possibly reflecting the role of the polyS domain in nuclear/nucleolar shuttling of Jmjd6.
Product Description
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-UBTF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution