Merck
  • Pulmonary T cell activation in response to chronic particulate air pollution.
Pulmonary T cell activation in response to chronic particulate air pollution.
American journal of physiology. Lung cellular and molecular physiology (2011-12-14)
Jeffrey A Deiuliis, Thomas Kampfrath, Jixin Zhong, Steve Oghumu, Andrei Maiseyeu, Lung Chi Chen, Qinghua Sun, Abhay R Satoskar, Sanjay Rajagopalan
ABSTRACT
The purpose of this study was to investigate the effects of chronically inhaled particulate matter <2.5 μm (PM(2.5)) on inflammatory cell populations in the lung and systemic circulation. A prominent component of air pollution exposure is a systemic inflammatory response that may exaggerate chronic diseases such as atherosclerosis and insulin resistance. T cell response was measured in wild-type C57B/L6, Foxp3-green fluorescent protein (GFP) "knockin," and chemokine receptor 3 knockout (CXCR3(-/-)) mice following 24-28 wk of PM(2.5) or filtered air. Chronic PM(2.5) exposure resulted in increased CXCR3-expressing CD4(+) and CD8(+) T cells in the lungs, spleen, and blood with elevation in CD11c(+) macrophages in the lung and oxidized derivatives of 1-palmitoyl-2-arachidonyl-sn-glycero-3-phosphorylcholine in wild-type mice. CXCR3 deficiency decreased T cells in the lung. GFP(+) regulatory T cells increased with PM(2.5) exposure in the spleen and blood of Foxp3-GFP mice but were present at very low levels in the lung irrespective of PM(2.5) exposure. Mixed lymphocyte cultures using primary, PM(2.5)-treated macrophages demonstrated enhanced T cell proliferation. Our experiments indicate that PM(2.5) potentiates a proinflammatory Th1 response involving increased homing of CXCR3(+) T effector cells to the lung and modulation of systemic T cell populations.
Sigma-Aldrich
Dispase® I, protease
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by trichloroacetic acid extraction