Merck
  • [Effect of phenazepam in various medicinal forms on psychophysical state of patients with anxiety disorders].
[Effect of phenazepam in various medicinal forms on psychophysical state of patients with anxiety disorders].
Eksperimental'naia i klinicheskaia farmakologiia (2011-08-04)
G G Neznamov, O A Dorofeeva, E S Teleshova, S A Siuniakov, L É Mametova
ABSTRACT
Differences between phenazepam formulations were found in the clinical study of drug effects on the psychophysiological state in patients with anxiety disorder. Phenazepam did not cause negative changes of psychophysiological parameters. In patients with anxiety disorders, the most significant positive therapeutic actionon psychophysiological state was observed in the case of using transdermal phenazepam formulation.
Product Description
Supelco
Phenazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®