Merck
  • Sudden cardiac arrest associated with use of a non-cardiac drug that reduces cardiac excitability: evidence from bench, bedside, and community.
Sudden cardiac arrest associated with use of a non-cardiac drug that reduces cardiac excitability: evidence from bench, bedside, and community.
European heart journal (2013-02-22)
Abdennasser Bardai, Ahmad S Amin, Marieke T Blom, Connie R Bezzina, Jocelyn Berdowski, Pim N J Langendijk, Leander Beekman, Christine A Klemens, Patrick C Souverein, Rudolph W Koster, Anthonius de Boer, Hanno L Tan
ABSTRACT
Non-cardiac drugs that impair cardiac repolarization (electrocardiographic QT prolongation) are associated with an increased sudden cardiac arrest (SCA) risk. Emerging evidence suggests that non-cardiac drugs that impair cardiac depolarization and excitability (electrocardiographic QRS prolongation) also increase the risk for SCA. Nortriptyline, which blocks the SCN5A-encoded cardiac sodium channel, may exemplify such drugs. We aimed to study whether nortriptyline increases the risk for SCA, and to establish the underlying mechanisms. We studied QRS durations during rest/exercise in an index patient who experienced ventricular tachycardia during exercise while using nortriptyline, and compared them with those of 55 controls with/without nortriptyline and 24 controls with Brugada syndrome (BrS) without nortriptyline, who carried an SCN5A mutation. We performed molecular-genetic (exon-trapping) and functional (patch-clamp) experiments to unravel the mechanisms of QRS prolongation by nortriptyline and the SCN5A mutation found in the index patient. We conducted a prospective community-based study among 944 victims of ECG-documented SCA and 4354-matched controls to determine the risk for SCA associated with nortriptyline use. Multiple mechanisms may act in concert to increase the risk for SCA during nortriptyline use. Pharmacological (nortriptyline), genetic (loss-of-function SCN5A mutation), and/or functional (sodium channel inactivation at fast heart rates) factors conspire to reduce the cardiac sodium current and increase the risk for SCA. Nortriptyline use in the community was associated with a 4.5-fold increase in the risk for SCA [adjusted OR: 4.5 (95% CI: 1.1-19.5)], particularly when other sodium channel-blocking factors were present. Nortriptyline increases the risk for SCA in the general population, particularly in the presence of genetic and/or non-genetic factors that decrease cardiac excitability by blocking the cardiac sodium channel.
Product Description
Supelco
Nortriptyline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Nortriptyline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nortriptyline hydrochloride, ≥98% (HPLC), powder