Merck
  • (-)-α-Bisabolol-induced gastroprotection is associated with reduction in lipid peroxidation, superoxide dismutase activity and neutrophil migration.
(-)-α-Bisabolol-induced gastroprotection is associated with reduction in lipid peroxidation, superoxide dismutase activity and neutrophil migration.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2011-09-20)
Nayrton Flávio Moura Rocha, Gersilene Valente de Oliveira, Fernanda Yvelize Ramos de Araújo, Emiliano Ricardo Vasconcelos Rios, Alyne Mara Rodrigues Carvalho, Leonardo Freire Vasconcelos, Danielle Silveira Macêdo, Pedro Marcos Gomes Soares, Damião Pergentino De Sousa, Francisca Cléa Florenço de Sousa
ABSTRACT
This work examined the gastroprotection of (-)-α-bisabolol, an unsaturated optically active sesquiterpene alcohol obtained by the direct distillation essential oil from plants. (-)-α-Bisabolol has been described as a compound capable of reducing the gastric ulcer area in response to absolute ethanol. We evaluated the gastroprotection of (-)-α-bisabolol in ethanol-induced gastric lesions model through histopathological assessment, measurement of the membrane lipids peroxidation (MDA), myeloperoxidase (MPO) activity, superoxide dismutase (SOD) activity, catalase (CAT) activity and the nitrite amount. Our results showed that (-)-α-bisabolol was able to reduce injuries associated with the administration of ethanol and the formation of thiobarbituric acid reactive substances (MDA) was also able to increase SOD activity and reduce the influx of cells inflammatory (neutrophils) in the gastric mucosa. The effect of (-)-α-bisabolol seems to be unrelated to the nitric oxide. (-)-α-Bisabolol caused a reduction of catalase activity. These findings show that (-)-α-bisabolol is able to decrease oxidative stress and inflammatory event associated with the lesions induced by ethanol.
Product Description
(−)-α-Bisabolol, primary reference standard
Supelco
(−)-α-Bisabolol, analytical standard
Sigma-Aldrich
(−)-α-Bisabolol, ≥93% (GC)