Merck
Cardioprotection through autophagy: ready for clinical trial?
Autophagy (2010-12-29)
Roberta A Gottlieb, Robert M Mentzer
ABSTRACT
Interventions that reduce infarct size in animal models have largely failed to improve outcome in patients suffering acute myocardial infarction (MI), or 'heart attack'. Our group recently reported a reduction of infarct size by chloramphenicol treatment in a porcine in vivo model of acute MI, through a mechanism involving the induction of autophagy. Since 2005 several studies have implicated autophagy as a target for cardioprotection.
Sigma-Aldrich
Chloramphenicol succinate sodium salt, ≥80% (HPLC)
Chloramphenicol sodium succinate, European Pharmacopoeia (EP) Reference Standard