Merck
[A convenient method of isoflavonone synthesis].
Bioorganicheskaia khimiia (2006-08-17)
A Aĭtmambetov, R T Tlegenov, A M Tokhtybaeva
ABSTRACT
Reaction of 2-hydroxydeoxybenzoins with bis(dimethylamino)methane in ethanol results in isoflavonones.
Sigma-Aldrich
4,4′-Methylenebis(N,N-dimethylaniline), 97.5% (GC)