Merck
  • Dysregulated anti-oxidant signalling and compromised mitochondrial integrity negatively influence regulatory T cell function and viability in liver disease.
Dysregulated anti-oxidant signalling and compromised mitochondrial integrity negatively influence regulatory T cell function and viability in liver disease.
EBioMedicine (2023-09-02)
Trishan Vaikunthanathan, Emmanuelle Landmann, Diana Marin Correa, Marco Romano, Silvia Cellone Trevelin, Qi Peng, Elena Crespo, Mauro Corrado, Juan-José Lozano, Erika L Pearce, Elena Perpinan, Anna Zoccarato, Leonard Siew, Joy Edwards-Hicks, Reenam Khan, Nguyet-Thin Luu, Mark R Thursz, Philip N Newsome, Marc Martinez-Llordella, Naina Shah, Robert I Lechler, Ajay M Shah, Alberto Sanchez-Fueyo, Giovanna Lombardi, Niloufar Safinia
ABSTRACT
Dysregulated inflammatory responses and oxidative stress are key pathogenic drivers of chronic inflammatory diseases such as liver cirrhosis (LC). Regulatory T cells (Tregs) are essential to prevent excessive immune activation and maintain tissue homeostasis. While inflammatory cues are well known to modulate the function and stability of Tregs, the extent to which Tregs are influenced by oxidative stress has not been fully explored. The phenotypic and functional properties of CD4+CD25+CD127lo/- Tregs isolated from patients with LC were compared to healthy controls (HC). Treg redox state was investigated by characterizing intracellular reactive oxygen species (ROS), NADPH oxidase-2 (Nox2) activity, mitochondrial function, morphology, and nuclear factor-erythroid 2-related factor (Nrf2) antioxidant signalling. The relevance of Nrf2 and its downstream target, Heme-oxygenase-1 (HO-1), in Treg function, stability, and survival, was further assessed using mouse models and CRISPR/Cas9-mediated HO-1 knock-out. Circulating Tregs from LC patients displayed a reduced suppressive function, correlating with liver disease severity, associated with phenotypic abnormalities and increased apoptosis. Mechanistically, this was linked to a dysregulated Nrf2 signalling with resultant lower levels of HO-1, enhanced Nox2 activation, and impaired mitochondrial respiration and integrity. The functional deficit in LC Tregs could be partially recapitulated by culturing control Tregs in patient sera. Our findings reveal that Tregs rely on functional redox homeostasis for their function, stability, and survival. Targeting Treg specific anti-oxidant pathways may have therapeutic potential to reverse the Treg impairment in conditions of oxidative damage such as advanced liver disease. This study was funded by the Wellcome Trust (211113/A/18/Z).
Sigma-Aldrich
Anti-phospho-p47 phox (pSer345) antibody produced in rabbit, affinity isolated antibody