Merck
  • Harnessing transcriptionally driven chromosomal instability adaptation to target therapy-refractory lethal prostate cancer.
Harnessing transcriptionally driven chromosomal instability adaptation to target therapy-refractory lethal prostate cancer.
Cell reports. Medicine (2023-02-15)
Brittiny Dhital, Sandra Santasusagna, Perumalraja Kirthika, Michael Xu, Peiyao Li, Marc Carceles-Cordon, Rajesh K Soni, Zhuoning Li, Ronald C Hendrickson, Matthew J Schiewer, William K Kelly, Cora N Sternberg, Jun Luo, Amaia Lujambio, Carlos Cordon-Cardo, Monica Alvarez-Fernandez, Marcos Malumbres, Haojie Huang, Adam Ertel, Josep Domingo-Domenech, Veronica Rodriguez-Bravo
ABSTRACT
Metastatic prostate cancer (PCa) inevitably acquires resistance to standard therapy preceding lethality. Here, we unveil a chromosomal instability (CIN) tolerance mechanism as a therapeutic vulnerability of therapy-refractory lethal PCa. Through genomic and transcriptomic analysis of patient datasets, we find that castration and chemotherapy-resistant tumors display the highest CIN and mitotic kinase levels. Functional genomics screening coupled with quantitative phosphoproteomics identify MASTL kinase as a survival vulnerability specific of chemotherapy-resistant PCa cells. Mechanistically, MASTL upregulation is driven by transcriptional rewiring mechanisms involving the non-canonical transcription factors androgen receptor splice variant 7 and E2F7 in a circuitry that restrains deleterious CIN and prevents cell death selectively in metastatic therapy-resistant PCa cells. Notably, MASTL pharmacological inhibition re-sensitizes tumors to standard therapy and improves survival of pre-clinical models. These results uncover a targetable mechanism promoting high CIN adaptation and survival of lethal PCa.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®