Merck
  • Evidence that spinal astrocytes but not microglia contribute to the pathogenesis of Paclitaxel-induced painful neuropathy.
Evidence that spinal astrocytes but not microglia contribute to the pathogenesis of Paclitaxel-induced painful neuropathy.
The journal of pain (2012-01-31)
Haijun Zhang, Seo-Yeon Yoon, Hongmei Zhang, Patrick M Dougherty
ABSTRACT
Paclitaxel often induces persistent painful neuropathy as its most common treatment-limiting side effect. Little is known concerning the underlying mechanisms. Given the prominent role of glial cells in many types of neuropathic pain, we investigated here the morphological and functional changes of spinal astrocytes and microglia in a rat model of paclitaxel-induced neuropathy. Immunohistochemistry, western blotting, and real-time polymerase chain reaction were performed with samples from 109 rats up to 28 days after paclitaxel treatment. Paclitaxel (2 mg/kg, i.p.) induced a rapid and persistent activation of spinal astrocytes assessed using glial fibrillary acidic protein, but not apparent activation of microglia assessed using OX42, Iba-1, and phosphorylated p38. In the context of astocyte activation, there was a significant downregulation of glial glutamate transporters GLAST and GLT-1 in spinal dorsal horn. The activation of spinal astrocytes by paclitaxel was not associated with expression of pro-inflammatory cytokines including tumor necrosis factor-α, interleukin-1β, or interleukin-6 in spinal dorsal horn. Systemic treatment with minocycline (50 mg/kg, i.p.) prevented activation of astrocytes and downregulation of glial glutamate transporters in spinal dorsal horn induced by paclitaxel. These data suggest the involvement of spinal astrocytes but not microglia in the pathogenesis of paclitaxel-induced neuropathy. Spinal astrocytes and/or glial glutamate transporters could be new therapeutic targets for paclitaxel-induced painful neuropathy.
Sigma-Aldrich
Anti-GLAST Antibody, clone 8C11.1, clone 8C11.1, from mouse
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)