Merck
  • Increase in Bioavailability of Nitric Oxide After Renal Denervation Improves Kidney Function in Sheep With Hypertensive Kidney Disease.
Increase in Bioavailability of Nitric Oxide After Renal Denervation Improves Kidney Function in Sheep With Hypertensive Kidney Disease.
Hypertension (Dallas, Tex. : 1979) (2021-03-02)
Reetu R Singh, Zoe M McArdle, Lindsea C Booth, Clive N May, Geoff A Head, Karen M Moritz, Markus P Schlaich, Kate M Denton
ABSTRACT
[Figure: see text].