Merck
  • Removal of Siloxanes from Model Biogas by Means of Deep Eutectic Solvents in Absorption Process.
Removal of Siloxanes from Model Biogas by Means of Deep Eutectic Solvents in Absorption Process.
Materials (Basel, Switzerland) (2021-01-10)
Edyta Słupek, Patrycja Makoś-Chełstowska, Jacek Gębicki
ABSTRACT
The paper presents the screening of 20 deep eutectic solvents (DESs) composed of tetrapropylammonium bromide (TPABr) and glycols in various molar ratios, and 6 conventional solvents as absorbents for removal of siloxanes from model biogas stream. The screening was achieved using the conductor-like screening model for real solvents (COSMO-RS) based on the comparison of siloxane solubility in DESs. For the DES which was characterized by the highest solubility of siloxanes, studies of physicochemical properties, i.e., viscosity, density, and melting point, were performed. DES composed of tetrapropylammonium bromide (TPABr) and tetraethylene glycol (TEG) in a 1:3 molar ratio was used as an absorbent in experimental studies in which several parameters were optimized, i.e., the temperature, absorbent volume, and model biogas flow rate. The mechanism of siloxanes removal was evaluated by means of an experimental FT-IR analysis as well as by theoretical studies based on σ-profile and σ-potential. On the basis of the obtained results, it can be concluded that TPABr:TEG (1:3) is a very effective absorption solvent for the removal of siloxanes from model biogas, and the main driving force of the absorption process is the formation of the hydrogen bonds between DES and siloxanes.
Sigma-Aldrich
Octamethyltrisiloxane, 98%
Sigma-Aldrich
Hexamethyldisiloxane, puriss., ≥98.5% (GC)