Merck
  • In Vivo Hydroxyl Radical Protein Footprinting for the Study of Protein Interactions in Caenorhabditis elegans.
In Vivo Hydroxyl Radical Protein Footprinting for the Study of Protein Interactions in Caenorhabditis elegans.
Journal of visualized experiments : JoVE (2020-04-21)
Jessica A Espino, Lisa M Jones
ABSTRACT
Fast oxidation of proteins (FPOP) is a hydroxyl radical protein footprinting (HRPF) method used to study protein structure, protein-ligand interactions, and protein-protein interactions. FPOP utilizes a KrF excimer laser at 248 nm for photolysis of hydrogen peroxide to generate hydroxyl radicals which in turn oxidatively modify solvent-accessible amino acid side chains. Recently, we expanded the use of FPOP of in vivo oxidative labeling in Caenorhabditis elegans (C. elegans), entitled IV-FPOP. The transparent nematodes have been used as model systems for many human diseases. Structural studies in C. elegans by IV-FPOP is feasible because of the animal's ability to uptake hydrogen peroxide, their transparency to laser irradiation at 248 nm, and the irreversible nature of the modification. The assembly of a microfluidic flow system for IV-FPOP labeling, IV-FPOP parameters, protein extraction, and LC-MS/MS optimized parameters are described herein.