Merck
  • A novel magnetic solid-phase extraction method for detection of 14 heterocyclic aromatic amines by UPLC-MS/MS in meat products.
A novel magnetic solid-phase extraction method for detection of 14 heterocyclic aromatic amines by UPLC-MS/MS in meat products.
Food chemistry (2020-08-14)
Shihui Shan, Yunjiao Ma, Changling Sun, Xiaolu Guo, Han Zheng, Xianbing Xu, Lei Qin, Jiangning Hu
ABSTRACT
The current study developed a cheap and effective method for the simultaneous extraction of 14 heterocyclic aromatic amines (HAAs) in food matrix. Core-shell Fe3O4@PDA nanoparticles were constructed and acted as the magnetic solid-phase extraction adsorbent to separate and purify HAAs from meat products for the first time. Then, UPLC-MS/MS technique was employed to identify and quantify the HAAs easily. Fe3O4@PDA nanoparticles were synthesized and characterized successfully. Totally 14 HAAs were completely separated in 19.99 min with good regression coefficients. LODs and LOQs were in the range of 0.013-0.247 ng/g and 0.056-0.803 ng/g, respectively. The intra-day precisions and inter-day precisions were below 9%. Except for IQ[4,5-b], Phe-p-1, PhIP, other 11 types of HAAs (DMIP, 1,5,6-TMIP, IQ, IQx, MeIQ, MeIQx, 7,8-DiMeIQx, AαC, MeAαC, Harman, Norharman) could acquire relatively high recoveries (71.06%-108.49%). The proposed method was successfully devoted to the evaluation of HAAs levels in 8 commercial meat products to verify the adaptability.