Merck
  • N,N'-Disuccinimidyl Carbonate: A Useful Reagent for Alkoxycarbonylation of Amines.
N,N'-Disuccinimidyl Carbonate: A Useful Reagent for Alkoxycarbonylation of Amines.
Tetrahedron letters (1992-05-12)
Arun K Ghosh, Tien T Duong, Sean P McKee, Wayne J Thompson
ABSTRACT
An efficient and mild method for alkoxycarbonylation of amines is described, utilizing commercially available N,N'-disuccinimidyl carbonate.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)