Merck
  • Oxidations of Benzhydrazide and Phenylacetic Hydrazide by Hexachloroiridate(IV): Reaction Mechanism and Structure-Reactivity Relationship.
Oxidations of Benzhydrazide and Phenylacetic Hydrazide by Hexachloroiridate(IV): Reaction Mechanism and Structure-Reactivity Relationship.
Molecules (Basel, Switzerland) (2020-01-17)
Xiaolai Zhang
ABSTRACT
Benz(o)hydrazide (BH) is the basic aryl hydrazide; aryl hydrazides have been pursued in the course of drug discovery. Oxidations of BH and phenylacetic hydrazide (PAH) by hexachloroiridate(IV) ([IrCl6]2-) were investigated by use of stopped-flow spectral, rapid spectral scan, RP-HPLC and NMR spectroscopic techniques. The oxidation reactions followed well-defined second-order kinetics and the observed second-order rate constant k' versus pH profiles were established over a wide pH range. Product analysis revealed that BH and PAH were cleanly oxidized to benzoic acid and phenylacetic acid, respectively. A reaction mechanism was proposed, resembling those suggested previously for the oxidations of isoniazid (INH) and nicotinic hydrazide (NH) by [IrCl6]2-. Rate constants of the rate-determining steps were evaluated, confirming a huge reactivity span of the protolysis species observed previously. The enolate species of BH is extremely reactive towards reduction of [IrCl6]2-. The determined middle-ranged negative values of activation entropies together with rapid scan spectra manifest that an outer-sphere electron transfer is probably taking place in the rate-determining steps. The reactivity of neutral species of hydrazides is clearly not correlated to the corresponding pKa values of the hydrazides. On the other hand, a linear correlation, logkenolate = (0.16 ± 0.07)pKenol + (6.1 ± 0.8), is found for the aryl hydrazides studied so far. The big intercept and the small slope of this correlation may pave a way for a rational design of new antioxidants based on aryl hydrazides. The present work also provides the pKa values for BH and PAH at 25.0 °C and 1.0 M ionic strength which were not reported before.
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium hexachloroiridate(IV) hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
2,4-Pyridinedicarboxylic acid, ≥98.0%