Merck
  • Voltammetric studies of hemoglobin-coated polystyrene latex bead films on pyrolytic graphite electrodes.
Voltammetric studies of hemoglobin-coated polystyrene latex bead films on pyrolytic graphite electrodes.
Biophysical chemistry (2004-07-02)
Hong Sun, Naifei Hu
ABSTRACT
A novel hemoglobin (Hb)-coated polystyrene (PS) latex bead film was deposited on pyrolytic graphite (PG) electrode surface. In the first step, positively charged Hb molecules in pH 5.0 buffers were adsorbed on the surface of negatively charged, 500 nm diameter PS latex beads bearing sulfate groups by electrostatic interaction. The aqueous dispersion of Hb-coated PS particles was then deposited on the surface of PG electrodes and, after evaporation of the solvent, Hb-PS films were formed. The Hb-PS film electrodes exhibited a pair of well-defined, quasi-reversible cyclic voltammetric (CV) peaks at about -0.36 V vs. SCE in pH 7.0 buffers, characteristic of Hb heme Fe(III)/Fe(II) redox couples. Positions of Soret absorption band of Hb-PS films suggest that Hb retains its near-native structure in the films in its dry form and in solution at medium pH. The Hb in PS films was also acted as a catalyst to catalyze electrochemical reduction of various substrates such as trichloroacetic acid (TCA), nitrite, oxygen and hydrogen peroxide.
Sigma-Aldrich
Latex beads, sulfate-modified polystyrene, fluorescent orange, aqueous suspension, 0.5 μm mean particle size
Sigma-Aldrich
Latex beads, sulfate-modified polystyrene, fluorescent orange, aqueous suspension, 0.1 μm mean particle size
SAFC
Latex beads, sulfate-modified polystyrene, fluorescent red, aqueous suspension, 0.1 μm mean particle size