- Boron-Boron Dehydrocoupling of Boranes Initiated by Reaction with Iodine.
Boron-Boron Dehydrocoupling of Boranes Initiated by Reaction with Iodine.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-03-02)
Jana Elias, Elisabeth Kaifer, Hans-Jörg Himmel
PMID30821401
ABSTRACT
A new boron-boron dehydrocoupling strategy was established, providing convenient access to some diborane(4) compounds starting from simple borane adducts under mild conditions. In contrast to the traditional pathway using a reducing reagent, the reduction from BIII to BII was paradoxically initiated by the addition of the oxidation-reagent iodine. A reaction pathway for this unusual reaction was proposed based on quantum-chemical calculations.