Merck
  • Kruppel-Like Factor 4 Regulates Granule Cell Pax6 Expression and Cell Proliferation in Early Cerebellar Development.
Kruppel-Like Factor 4 Regulates Granule Cell Pax6 Expression and Cell Proliferation in Early Cerebellar Development.
PloS one (2015-08-01)
Peter Zhang, Thomas Ha, Matt Larouche, Douglas Swanson, Dan Goldowitz
ABSTRACT
Kruppel-like factor 4 (Klf4) is a transcription factor that regulates many important cellular processes in stem cell biology, cancer, and development. We used histological and molecular methods to study the expression of Klf4 in embryonic development of the normal and Klf4 knockout cerebellum. We find that Klf4 is expressed strongly in early granule cell progenitor development but tails-off considerably by the end of embryonic development. Klf4 is also co-expressed with Pax6 in these cells. In the Klf4-null mouse, which is perinatal lethal, Klf4 positively regulates Pax6 expression and regulates the proliferation of neuronal progenitors in the rhombic lip, external granular layer and the neuroepithelium. This paper is the first to describe a role for Klf4 in the cerebellum and provides insight into this gene's function in neuronal development.
Sigma-Aldrich
Anti-GLAST Antibody, clone 8C11.1, clone 8C11.1, from mouse
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.