Merck
  • High-performance and scalable metal-chalcogenide semiconductors and devices via chalco-gel routes.
High-performance and scalable metal-chalcogenide semiconductors and devices via chalco-gel routes.
Science advances (2018-04-18)
Sung Min Kwon, Jong Kook Won, Jeong-Wan Jo, Jaehyun Kim, Hee-Joong Kim, Hyuck-In Kwon, Jaekyun Kim, Sangdoo Ahn, Yong-Hoon Kim, Myoung-Jae Lee, Hyung-Ik Lee, Tobin J Marks, Myung-Gil Kim, Sung Kyu Park
ABSTRACT
We report a general strategy for obtaining high-quality, large-area metal-chalcogenide semiconductor films from precursors combining chelated metal salts with chalcoureas or chalcoamides. Using conventional organic solvents, such precursors enable the expeditious formation of chalco-gels, which are easily transformed into the corresponding high-performance metal-chalcogenide thin films with large, uniform areas. Diverse metal chalcogenides and their alloys (MQ x : M = Zn, Cd, In, Sb, Pb; Q = S, Se, Te) are successfully synthesized at relatively low processing temperatures (<400°C). The versatility of this scalable route is demonstrated by the fabrication of large-area thin-film transistors (TFTs), optoelectronic devices, and integrated circuits on a 4-inch Si wafer and 2.5-inch borosilicate glass substrates in ambient air using CdS, CdSe, and In2Se3 active layers. The CdSe TFTs exhibit a maximum field-effect mobility greater than 300 cm2 V-1 s-1 with an on/off current ratio of >107 and good operational stability (threshold voltage shift < 0.5 V at a positive gate bias stress of 10 ks). In addition, metal chalcogenide-based phototransistors with a photodetectivity of >1013 Jones and seven-stage ring oscillators operating at a speed of ~2.6 MHz (propagation delay of < 27 ns per stage) are demonstrated.
Sigma-Aldrich
Selenourea, 98%