
Venlafaxine for system suitability

Synonym(s):
Venlafaxine hydrochloride, (+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, Effexor
Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing