
TDA Reagent

Synonym(s):
Iron(III) chloride solution, Ferric chloride solution
Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing