
Selenomethionine

Synonym(s):
Seleno-DL-methionine
Empirical Formula (Hill Notation):
C5H11NO2Se
CAS Number:
Molecular Weight:
196.11
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1