
Selegiline Related Compound D

Synonym(s):
R-(–)-Desmethyldeprenyl hydrochloride, (R)-N-(1-Phenylpropan-2-yl)prop-2-yn-1-amine hydrochloride, N-Desmethylselegiline hydrochloride, L-Nordeprenyl hydrochloride, Norselegiline hydrochloride
Empirical Formula (Hill Notation):
C12H15N · HCl
CAS Number:
Molecular Weight:
209.72
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing