
Repaglinide impurity E

Synonym(s):
2-Ethoxy-4-{2-{{(1R)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl}amino}-2-oxoethyl}benzoic acid, (R)-(-)-Repaglinide
Empirical Formula (Hill Notation):
C27H36N2O4
CAS Number:
Molecular Weight:
452.59
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing