
Phenolphthalein β-D-glucuronide

Synonym(s):
Phenolphthalein β-D-glucosiduronic acid, Phenolphthalein glucuronic acid, Phenolphthalein mono-β-D-glucosiduronic acid
Empirical Formula (Hill Notation):
C26H22O10
CAS Number:
Molecular Weight:
494.45
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing