
Paroxetine Related Compound B

Synonym(s):
trans-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine hydrochloride, (3S,4R)-3-[(Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine hydrochloride hydrochloride
Empirical Formula (Hill Notation):
C19H21NO3 · HCl
Molecular Weight:
347.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing