
Histidine

Synonym(s):
L-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773
Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing