
Glycitein

Synonym(s):
4′,7-Dihydroxy-6-methoxyisoflavone, Glycetein
Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Products
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4