Skip to Content
Merck
Disopyramide phosphate salt

Disopyramide phosphate salt

Disopyramide phosphate salt
Synonym(s):
α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide
Linear Formula:
C21H29N3O · H3PO4
CAS Number:
Molecular Weight:
437.47
EC Number:

Products

Product Category
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing