
Coenzyme Q9

Synonym(s):
Q-9, Ubiquinone-45, Ubiquinone-9
Empirical Formula (Hill Notation):
C54H82O4
CAS Number:
Molecular Weight:
795.23
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing