Atazanavir Related Compound A
Synonym(s):
(2S)-2-[(Methoxycarbonyl)amino]-3,3-dimethylbutanoic acid, N-Methoxycarbonyl-3-methyl-L-valine
Empirical Formula (Hill Notation):
C8H15NO4
CAS Number:
Molecular Weight:
189.21
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing