
Apocarotenal

Synonym(s):
trans-β-Apo-8′-carotenal, Apocarotenal
Empirical Formula (Hill Notation):
C30H40O
CAS Number:
Molecular Weight:
416.64
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing