
α-Carotene

Synonym(s):
(+)-α-Carotene
Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing