
6-Formylpterin

Synonym(s):
2-Amino-4-hydroxy-6-formylpteridine, 2-Amino-6-formylpteridin-4-one, Pterin-6-aldehyde
Empirical Formula (Hill Notation):
C7H5N5O2
CAS Number:
Molecular Weight:
191.15
Products
Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing