
1,10-Phenanthroline monohydrate

Synonym(s):
o-Phenanthroline monohydrate
Empirical Formula (Hill Notation):
C12H8N2 · H2O
CAS Number:
Molecular Weight:
198.22
Products
Product Category
Biological Source
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Package Size
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing