Human Serum for Cell Culture
![Cell Culture Supplements & Reagents A pipette dropping a clear liquid into a small glass dish, with a blurred bottle labeled with orange text in the background, set on a reflective surface.](/deepweb/assets/sigmaaldrich/marketing/global/images/categories/cell-analysis/human-serum/human-serum.jpg)
Serum is the fluid that remains once clotting factors are removed from blood plasma. A common supplement in cell culture media, serum provides a mix of hormones, growth and attachment factors, buffering agents, and other nutritional components to cells. Some studies have determined that human serum is better suited than FBS for certain human cell culture applications, such as cell invasion studies and the formation of spheroids. Our human serum for cell culture is tested for the presence of viruses, bacteria, hemoglobin, and toxins, and then serially filtered under sterile conditions.
Related Product Resources
- Download: Cell Culture Manual
Cultivating healthy cell growth means depending on a quality mix of media, sera, and reagents. That's why we offer an unparalleled line of cell culture solutions. From classical and specialty media, to fetal bovine serum, supplements and reagents, our offering is extensive and comprehensive.
- Article: Fetal Bovine Serum that Makes the Grade
Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.
- Article: Fetal Bovine Serum (FBS) - What You Need to Know
Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.
- Article: Serum for Cell and Tissue Culture FAQs
Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.
Human AB Serum Products
Human adult AB serum is used for culturing human cell lines, including human stem cell lines. We offer both AB off-the-clot and AB plasma products. Human AB serum is prepared from whole blood that is allowed to clot naturally, called OTC or Off-The-Clot serum. Human serum from clotted whole blood is used most frequently however, as it contains more growth factors for cell growth.
AB human serum can also be plasma-derived or prepared from whole blood that is not allowed to coagulate. Once the plasma is separated from whole blood, it is treated with calcium to activate the clotting cascade and centrifuged to extract the serum. This preparation is used when testing sera for growth factors.
We offer heat inactivated human AB serum both from plasma and clotted whole blood. Adult serum contains complement, a component in sera that can be harmful to some cells or have deleterious effects in downstream immunoassays. Heat inactivation of serum is inactivates complement, and this serum is mainly used when culturing hematopoietic cells or when performing immunological assays, such as mixed-lymphocyte reactions or neutralization assays.
Human Serum from Platelet Poor Plasma
Human serum from platelet poor plasma (PPP) is a media supplement that is often used in studies that look at the effects that platelet lysates have on the cellular growth, such as on human skin fibroblasts and articular chondrocytes growth. PPP human sera is also used when differentiating human induced pluripotent stem cells (hiPS) to hiPS-brain microvascular endothelial cells or in platelet derived growth factor (PDGF) purification assays.
Applications for Human Serum
Human serum from blood type AB donors lacks antibodies specific for both A and B blood-type antigens, making it suitable for transplantation and cell therapy. Other biotechnical and biopharmaceutical applications include tissue engineering, the expansion of mesenchymal stem cells from adipose tissue, or of mesenchymal stromal cells from human bone marrow. In addition, AB serum is essential for expanding peripheral blood-derived NK cells, and for enhancing pre-transplant human islet culture.