Skip to Content
Merck
All Photos(1)

Key Documents

CRM45995

Supelco

Cartap hydrochloride

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N3O2S2 · HCl
CAS Number:
Molecular Weight:
273.80
Beilstein:
5157539
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)C(CSC(N)=O)CSC(N)=O

InChI

1S/C7H15N3O2S2.ClH/c1-10(2)5(3-13-6(8)11)4-14-7(9)12;/h5H,3-4H2,1-2H3,(H2,8,11)(H2,9,12);1H

InChI key

MSHXTAQSSIEBQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Cartap hydrochloride is a nereistoxin analogue insecticide used to control chewing and sucking pests on soya beans, peanuts, maize, sugarbeet, wheat and several fruits like apples, pears and plums. Under Reg. (EC) No 1107/2009 cartap hydrochloride is not approved for use in the European Union as a plant protection product. Maximum residue Cartap hydrochloride is a nereistoxin analogue insecticide used to control chewing and sucking pests on soya beans, peanuts, maize, sugarbeet, wheat and several fruits like apples, pears and plums. Under Reg. (EC) No 1107/2009 cartap hydrochloride is not approved for use in the European Union as a plant protection product. Maximum residue levels (MRLs) for some products of plant origin have been set according to Reg. (EU) 149/2008 for cartap hydrochloride to 0.1 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Environmental

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service