Skip to Content
Merck

SML1350

EPPTB

≥95% (HPLC)

Synonym(s):

N-(3-Ethoxy-phenyl)-4-pyrrolidin-1-yl-3-trifluoromethyl-benzamide, RO5212773

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H21F3N2O2
CAS Number:
Molecular Weight:
378.39
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCOC1=CC=CC(NC(C2=CC(C(F)(F)F)=C(N3CCCC3)C=C2)=O)=C1

InChI

1S/C20H21F3N2O2/c1-2-27-16-7-5-6-15(13-16)24-19(26)14-8-9-18(25-10-3-4-11-25)17(12-14)20(21,22)23/h5-9,12-13H,2-4,10-11H2,1H3,(H,24,26)

InChI key

KLFVWQCQUXXLOU-UHFFFAOYSA-N

Biochem/physiol Actions

EPPTB is a selective antagonist of the trace amine-associated receptor 1 (TAAR1), a G protein-coupled receptor (GPCR) activated by endogenous trace amines (tyramine, tryptamine, synephrine, octopamine, β)-phenylethylamine), which are chemically similar to monoaminergic neurotransmitters (epinephrine, norepinephrine, dopamine). TAAR1 is considered a promising target for the treatment of neuropsychiatric disorders, however study of TAAR1 has hampered by the fact that selective ligands have not been available. EPPTB is the first potent and selective TAAR1 antagonist with an IC50 of 28 nM at mouse TAAR1. EPPTB is less selective for rat and human with IC50 values of 4539 and 7487 nM, respectively.
EPPTB is a selective antagonist of the trace amine-associated receptor 1 (TAAR1).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library