Skip to Content
Merck

MAK477

Calcium Assay Kit

sufficient for 500 colorimetric tests

Synonym(s):

Calcium Quantitative Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

application(s)

pharmaceutical

detection method

colorimetric

relevant disease(s)

musculoskeletal disorders; genitourinary disorders; cancer; endocrinological disorders

shipped in

ambient

storage temp.

2-8°C

General description

Calcium is measured to monitor diseases of the bone or calcium regulation disorders. Increased calcium levels in serum are reported in hyperparathyroidism, metastatic bone lesions and hypervitaminosis. While decreased levels are observed in hypoparathyroidism, nephrosis, rickets, steatorrhea, nephritis and calcium-losing syndromes. Urinary calcium levels aid in understanding how the kidneys handle calcium in certain diseases of the parathyroid gland. Urinary calcium levels are also essential in the evaluation of kidney stones.

Application

The kit is suitable for calcium determination in blood, urine, saliva and other biological samples, as well as for studying the effects of drugs on calcium metabolism.

Biochem/physiol Actions

This Calcium Assay Kit is designed to measure calcium directly in biological samples without any pre-treatment. Phenolsulphonephthalein dye in the kit forms a very stable blue colored complex specifically with free calcium. The intensity of the color, measured at 612 nm, is directly proportional to the calcium concentration in the sample. The optimized formulation minimizes any interference by substances such as magnesium, lipids, protein, and bilirubin.

Features and Benefits

  • The linear detection range of the kit is 0.08 mg/dL (20 µM) to 20 mg/dL (5 mM).
  • Compatible with high-throughput handling systems.
  • The optimized formulation has greatly enhanced reagent and signal stability.
  • Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples i.e., in the presence of lipids, protein, magnesium, iron and zinc.


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library