ERMBF426D
Soya 305423 GMO standard ERM-BF426
ERM®, certified reference material, level 3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
ERM®
composition
Soya 305423, 10%
manufacturer/tradename
JRC
application(s)
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Analysis Note
For more information please see:
ERMBF426D
ERMBF426D
Legal Information
ERM is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service