Skip to Content
Merck
All Photos(1)

Key Documents

BCR153R

Dibenz[a,h]acridine

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13N
CAS Number:
Molecular Weight:
279.33
Beilstein:
209261
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3nc4c(ccc5ccccc45)cc23

InChI

1S/C21H13N/c1-3-7-17-14(5-1)11-12-20-19(17)13-16-10-9-15-6-2-4-8-18(15)21(16)22-20/h1-13H

InChI key

JNCSIWAONQTVCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR153R

Legal Information

BCR is a registered trademark of European Commission

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service