Skip to Content
Merck
All Photos(1)

Key Documents

HSTCMAG28PMX13BK

Millipore

MILLIPLEX® Human High Sensitivity T Cell Panel Premixed 13-plex - Space Saver (Bulk) Packaging

Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

sensitivity: 0.11-8.17 pg/mL
(Overnight Incubation minDC)

standard curve range: 0.18-750 pg/mL
(IL-6)

standard curve range: 0.24-1,000 pg/mL
(IL-13)

standard curve range: 0.31-1,250 pg/mL
(IL-8)

standard curve range: 0.37-1,500 pg/mL
(IL-7)

standard curve range: 0.45-1,750 pg/mL
(TNFα)

standard curve range: 0.49-2,000 pg/mL
(IL-1β)

standard curve range: 0.49-2,000 pg/mL
(IL-12 (p70))

standard curve range: 0.49-2,000 pg/mL
(IL-2)

standard curve range: 0.49-2,000 pg/mL
(IL-5)

standard curve range: 0.61-2,500 pg/mL
(IFNγ)

standard curve range: 1.22-5,000 pg/mL
(GM-CSF)

standard curve range: 1.46-6,000 pg/mL
(IL-10)

standard curve range: 1.83-7,500 pg/mL
(IL-4)

inter-assay cv: <15%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cytokines are immunomodulatory polypeptides that play key roles in both adaptive and innate immune responses. A generic term, “cytokines” includes myokines (produced by muscle cells), lymphokines (produced by activated Th cells), interleukins (acting as mediators between T cells) and chemokines (responsible for T-cell migration). One of the regulatory mechanisms of the immune system, cytokines act at the recognition, activation, or effector phases of an immune response, modulating the development and functional activities of the subtypes of T cells, B cells and myeloid cells. Consequently, research involving cytokines plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens, especially those responses that make up the inflammatory process. Low levels of inflammation are involved in many clinical and sub-clinical disease states, such as autoimmune disease, cardiovascular disease, diabetes, neurological disorders, and cancer. Measuring picogram levels of cytokines, therefore, is critical for understanding the pathogenesis of these diseases.

The MILLIPLEX® Human High Sensitivity T Cell Magnetic Bead Panel is to be used for the simultaneous quantification of any or all of the following analytes in human plasma, serum, and cell/tissue culture supernatant samples: Fractalkine, GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-13, IL-17A, IL-21, IL-23, ITAC, MIP-1α, MIP-1β, MIP-3α
and TNFα.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Application

  • Analytes: GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-13, TNF-α
  • Because there are unique features concerning sample prep and assay set-up, please read the entire protocol before running the assay.
  • Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology
  • Research Sub-Category: Immune Response, Metabolic Disorders, Chronic Inflammatory Disease, Cardiovascular Disease, Neuroscience

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: Please see kit protocol for individual assay sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sascha N Goonewardena et al.
Cardiovascular drugs and therapy (2021-11-19)
Vascular and immune dysfunction are hallmarks of severe acute respiratory syndrome coronavirus-2 (SARS-CoV-2) infections and coronavirus disease 2019 (COVID-19). Although our understanding of the pathogenesis of COVID-19 has rapidly evolved, much of the focus has been on the immune mechanisms
Jeremias Bayon et al.
Cardiology research, 11(5), 319-327 (2020-08-28)
Cyclophilins (Cyps) are a family of peptidyl-prolyl cis/trans isomerases consistently involved in cardiovascular diseases through the inflammation pathway. This study aims to investigate the serum levels of Cyps (CypA, CypB, CypC and CypD) in patients with coronary artery disease (CAD)
Amparo Alfonso et al.
Scientific reports, 9(1), 10576-10576 (2019-07-25)
Cyclophilins (Cyps) are ubiquitous proteins that belong to the immunophilins family consistently associated with inflammatory and cardiovascular diseases. While levels of CypA have been extensively studied, less data are available for other Cyps. The purpose of this case-control study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service