Skip to Content
Merck
All Photos(1)

Key Documents

8.21189

Sigma-Aldrich

1,3,5-Trioxane

for synthesis

Synonym(s):

1,3,5-Trioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-812-5
NACRES:
NA.22

vapor pressure

7.5 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

410 °C

potency

8190 mg/kg LD50, oral (Rat)
>3980 mg/kg LD50, skin (Rabbit)

expl. lim.

3.6-28.7 % (v/v)

bp

114-115 °C/1013 hPa

mp

60-62 °C

transition temp

flash point 45 °C

density

1.17 g/cm3 at 65 °C

storage temp.

2-30°C

InChI

1S/C3H6O3/c1-4-2-6-3-5-1/h1-3H2

InChI key

BGJSXRVXTHVRSN-UHFFFAOYSA-N

Application

  • Organic Salts and 1,3,5-Trioxane Synthesis: Discusses the effect of various organic salts on the synthesis of 1,3,5-Trioxane, indicating significant influences on yield due to the salts′ properties (Yin et al., 2016).
  • Separation Technologies: Explores the use of imidazolium-based polyionic liquids for separating the 1,3,5-Trioxane-Water/Ethanol-Water system, based on experimental verification and molecular simulations (Qu et al., 2022).
  • Production from Dimethoxymethane: Studies polyoxymethylene dimethyl ethers production from dimethoxymethane and 1,3,5-Trioxane over SO42-/ZrO2-TiO2, providing insights into efficient production processes (Yao et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 60 °C
Melting range (upper value): ≤ 63 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Repr. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service