Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
SNAP-N-SPIKE®/SNAP-N-SHOOT®
packaging
ampule of 3 × 1.2 mL
manufacturer/tradename
Cerilliant®
concentration
in H2O (varied conc.)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
multi-component solution
storage temp.
2-8°C
General description
Application
Analysis Note
C1 C2 C3
Acetone 500 1000 4000
Ethanol 500 1000 4000
Isopropanol 5001000 4000
Methanol5001000 4000
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| format multi-component solution | format multi-component solution | format multi-component solution | format multi-component solution |
| form liquid | form liquid | form liquid | form liquid |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A-054-3X1.2ML | 04061833326152 |



