Skip to Content
Merck
All Photos(2)

Key Documents

860371P

Avanti

14:0 PE-d54

Avanti Research - A Croda Brand 860371P, powder

Synonym(s):

1,2-dimyristoyl-d54-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H12NO8PD54
CAS Number:
Molecular Weight:
690.19
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860371P-10mg)
pkg of 1 × 100 mg (860371P-100mg)
pkg of 1 × 25 mg (860371P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860371P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H]

InChI

1S/C33H66NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-32(35)39-29-31(30-41-43(37,38)40-28-27-34)42-33(36)26-24-22-20-18-16-14-12-10-8-6-4-2/h31H,3-30,34H2,1-2H3,(H,37,38)/t31-/m1/s1/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2

InChI key

NEZDNQCXEZDCBI-QHDWWDETSA-N

General description

1,2-dimyristoyl-d54-sn-glycero-3-phosphoethanolamine (14:0 PE-d54) is a deuterated phosphoethanolamine (PE) in which 54 protons of dimyristoyl are replaced by deuterium.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Packaging

5 mL Amber Glass Screw Cap Vial (860371P-100mg)
5 mL Amber Glass Screw Cap Vial (860371P-10mg)
5 mL Amber Glass Screw Cap Vial (860371P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of materials chemistry. B (2012)
Nicole Harmouche et al.
Biophysical journal, 115(6), 1033-1044 (2018-09-10)
A synergistic enhancement of activities has been described for the amphipathic cationic antimicrobial peptides magainin 2 and PGLa when tested in antimicrobial assays or in biophysical experiments using model membranes. In the presence of magainin 2, PGLa changes from an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service