Merck
  • Synthesis of quinolines, 2-quinolones, phenanthridines, and 6(5h)-phenanthridinones via palladium[0]-mediated Ullmann cross-coupling of 1-bromo-2-nitroarenes with beta-halo-enals, -enones, or -esters.
Synthesis of quinolines, 2-quinolones, phenanthridines, and 6(5h)-phenanthridinones via palladium[0]-mediated Ullmann cross-coupling of 1-bromo-2-nitroarenes with beta-halo-enals, -enones, or -esters.
Organic letters (2004-07-30)
Martin G Banwell, David W Lupton, Xinghua Ma, Jens Renner, Magne O Sydnes
ABSTRACT
Palladium[0]-mediated Ullmann cross-coupling of 1-bromo-2-nitrobenzene (1 R = H) and its derivatives with a range of beta-halo-enals, -enones, or -esters readily affords the corresponding beta-aryl derivatives, which are converted into the corresponding quinolines, 2-quinolones, phenanthridines, or 6(5H)-phenanthridinones on reaction with dihydrogen in the presence of Pd on C or with TiCl(3) in aqueous acetone. [reaction: see text]
Sigma-Aldrich
1-Bromo-2-nitrobenzene, 98%